The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139025.4 | NP_620594.1 | 17 - 18 | 2543 | 114 |
NM_139026.4 | NP_620595.1 | 17 - 18 | 2450 | 114 | |
NM_139027.4 | NP_620596.2 | 17 - 18 | 2543 | 114 | |
XM_011518176.2 | XP_011516478.1 | 10 - 11 | 1195 | 114 | |
XM_011518178.1 | XP_011516480.1 | 7 - 8 | 869 | 114 | |
XM_011518179.1 | XP_011516481.1 | 7 - 8 | 912 | 114 | |
XM_017014232.1 | XP_016869721.1 | 17 - 18 | 2754 | 114 | |
XM_017014233.1 | XP_016869722.1 | 17 - 18 | 2155 | 114 | |
XM_017014234.1 | XP_016869723.1 | 11 - 12 | 1343 | 114 | |
XM_017014235.1 | XP_016869724.1 | 17 - 18 | 3324 | 114 | |
GenBank mRNA | AB069698.2 | - | 17 - 18 | 2543 | 114 |
AF414401.1 | - | 17 - 18 | 2160 | 114 | |
AJ305314.1 | - | 17 - 18 | 2126 | 114 | |
AJ420810.1 | - | 17 - 18 | 2219 | 114 | |
AY055376.1 | - | 17 - 18 | 2207 | 114 | |
AY358118.1 | - | 10 - 11 | 1311 | 114 | |
BC103830.1 | - | 10 - 11 | 1304 | 114 |