The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139025.4 | NP_620594.1 | 24 - 25 | 3694 | 88 |
NM_139026.4 | NP_620595.1 | 24 - 25 | 3601 | 88 | |
NM_139027.4 | NP_620596.2 | 24 - 25 | 3694 | 88 | |
NR_024514.2 | 14 - 15 | 2085 | 88 | ||
XM_011518176.2 | XP_011516478.1 | 17 - 18 | 2346 | 88 | |
XM_011518178.1 | XP_011516480.1 | 14 - 15 | 2020 | 88 | |
XM_011518179.1 | XP_011516481.1 | 14 - 15 | 2063 | 88 | |
XM_017014232.1 | XP_016869721.1 | 24 - 25 | 3905 | 88 | |
XM_017014233.1 | XP_016869722.1 | 24 - 25 | 3306 | 88 | |
XM_017014234.1 | XP_016869723.1 | 18 - 19 | 2494 | 88 | |
GenBank mRNA | AB069698.2 | - | 24 - 25 | 3694 | 88 |
AF414401.1 | - | 24 - 25 | 3311 | 88 | |
AJ305314.1 | - | 24 - 25 | 3277 | 88 | |
AJ420810.1 | - | 24 - 25 | 3370 | 88 | |
AK124983.1 | - | 2 - 3 | 271 | 88 | |
AY055376.1 | - | 24 - 25 | 3358 | 88 | |
AY358118.1 | - | 17 - 18 | 2462 | 88 |