Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
Chromosome Location
Chr.11: 117205071 - 117232525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004716.3 NP_004707.2 9 - 10 1787 102
XM_006718940.3 XP_006719003.1 2 - 3 546 102
XM_017018546.1 XP_016874035.1 4 - 5 315 102
GenBank mRNA AK097207.1 - 1 - 2 962 102
AK222907.1 - 9 - 10 1327 102
AK312429.1 - 9 - 10 1362 102
BC006357.2 - 9 - 10 1305 102
BC010696.1 - 9 - 10 1357 102
BT006870.1 - 7 - 8 1156 102
U33849.1 - 9 - 10 1307 102
U40623.1 - 7 - 8 1177 102

View Full Product Details