The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012194.2 | NP_036326.2 | 20 - 20 | 7638 | 127 |
XM_005252847.3 | XP_005252904.2 | 22 - 22 | 7732 | 127 | |
XM_005252848.3 | XP_005252905.1 | 20 - 20 | 7653 | 127 | |
XM_011519970.2 | XP_011518272.1 | 21 - 21 | 6298 | 127 | |
XM_017017484.1 | XP_016872973.1 | 20 - 20 | 7656 | 127 | |
XM_017017486.1 | XP_016872975.1 | 21 - 21 | 6295 | 127 |