The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152744.3 | NP_689957.3 | 9 - 10 | 1565 | 95 |
XM_011515188.1 | XP_011513490.1 | 3 - 4 | 403 | 95 | |
XM_011515189.1 | XP_011513491.1 | 3 - 4 | 403 | 95 | |
XM_011515190.1 | XP_011513492.1 | 3 - 4 | 403 | 95 | |
XM_017011837.1 | XP_016867326.1 | 3 - 4 | 403 | 95 | |
XM_017011838.1 | XP_016867327.1 | 3 - 4 | 403 | 95 | |
XM_017011839.1 | XP_016867328.1 | 3 - 4 | 403 | 95 | |
XM_017011840.1 | XP_016867329.1 | 3 - 4 | 403 | 95 | |
GenBank mRNA | AY310398.1 | - | 9 - 10 | 1565 | 95 |