The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002832.2 | - | 87 | ||
NR_033312.1 | - | 87 | |||
NR_033313.1 | - | 87 | |||
NR_033314.1 | - | 87 | |||
NR_033315.1 | - | 87 | |||
GenBank mRNA | AY054391.4 | - | - | 87 | |
AY054401.3 | - | - | 87 | ||
AY054402.3 | - | - | 87 | ||
AY054403.4 | - | - | 87 | ||
AY054404.4 | - | - | 87 | ||
AY054405.4 | - | - | 87 | ||
AY513486.3 | - | - | 87 | ||
EF689023.1 | - | - | 87 | ||
EF689024.1 | - | - | 87 | ||
EF689025.1 | - | - | 87 | ||
EF689026.1 | - | - | 87 | ||
EF689027.1 | - | - | 87 | ||
EF689028.1 | - | - | 87 | ||
EF689029.1 | - | - | 87 | ||
EF689030.1 | - | - | 87 | ||
EF689031.1 | - | - | 87 | ||
EF689032.1 | - | - | 87 | ||
EF689033.1 | - | - | 87 | ||
EF689034.1 | - | - | 87 | ||
EF689035.1 | - | - | 87 | ||
EF689036.1 | - | - | 87 | ||
EF689037.1 | - | - | 87 | ||
EF689038.1 | - | - | 87 | ||
EF689039.1 | - | - | 87 | ||
EF689040.1 | - | - | 87 | ||
EF689041.1 | - | - | 87 | ||
EF689042.1 | - | - | 87 |