The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033314.1 | 5 - 6 | 597 | 116 | |
NR_033315.1 | 4 - 5 | 528 | 116 | ||
GenBank mRNA | AY054405.4 | - | 5 - 6 | 627 | 116 |
AY513486.3 | - | 4 - 5 | 528 | 116 | |
EF689029.1 | - | 3 - 4 | 436 | 116 | |
EF689035.1 | - | 5 - 6 | 602 | 116 | |
EF689041.1 | - | 4 - 5 | 533 | 116 | |
EF689042.1 | - | 5 - 6 | 602 | 116 |