Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMC5
Entrez Gene ID
79838
Gene Name
transmembrane channel like 5
Gene Aliases
-
Chromosome Location
Chr.16: 19410539 - 19499113 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105248.1 NP_001098718.1 8 - 9 2228 117
NM_001105249.1 NP_001098719.1 8 - 9 2228 117
NM_001261841.1 NP_001248770.1 8 - 9 2183 117
NM_001308161.1 NP_001295090.1 8 - 9 2021 117
NM_024780.4 NP_079056.2 4 - 5 998 117
GenBank mRNA AK097718.1 - 4 - 5 963 117
AK131202.1 - 5 - 6 718 117
AK299055.1 - 8 - 9 2021 117
AK304431.1 - 8 - 9 2183 117
AY236494.1 - 4 - 5 998 117
AY263164.1 - 4 - 5 741 117
AY358155.1 - 6 - 7 1577 117
BC027602.1 - 4 - 5 953 117
BC038118.2 - 4 - 5 693 117
CR749359.1 - 6 - 7 883 117

View Full Product Details