The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014983.2 | NP_055798.2 | 13 - 14 | 2984 | 60 |
XM_006714765.2 | XP_006714828.1 | 12 - 13 | 2486 | 60 | |
GenBank mRNA | BC005880.2 | - | 7 - 8 | 988 | 60 |
BC051025.1 | - | 13 - 14 | 2850 | 60 | |
BC092520.1 | - | 13 - 14 | 2857 | 60 | |
D83778.1 | - | 13 - 14 | 2839 | 60 |