Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGXB3
Entrez Gene ID
22993
Gene Name
HMG-box containing 3
Gene Aliases
HMGX3, SMF
Chromosome Location
Chr.5: 150000606 - 150053143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014983.2 NP_055798.2 15 - 16 3300 86
XM_006714765.2 XP_006714828.1 14 - 15 2802 86
GenBank mRNA BC005880.2 - 9 - 10 1304 86
BC051025.1 - 15 - 16 3166 86
BC063691.1 - 1 - 2 68 86
BC092520.1 - 15 - 16 3173 86
D83778.1 - 15 - 16 3155 86

View Full Product Details