The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014983.2 | NP_055798.2 | 2 - 3 | 714 | 142 |
XM_006714765.2 | XP_006714828.1 | 2 - 3 | 714 | 142 | |
GenBank mRNA | BC051025.1 | - | 2 - 3 | 580 | 142 |
BC092520.1 | - | 2 - 3 | 587 | 142 | |
D83778.1 | - | 2 - 3 | 569 | 142 |