Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
Chromosome Location
Chr.15: 45092653 - 45114161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014080.4 NP_054799.4 19 - 20 2764 95
XM_005254421.2 XP_005254478.1 19 - 20 2764 95
GenBank mRNA AB209010.1 - 13 - 14 4292 95
AF181972.1 - 11 - 12 1547 95
AF230496.1 - 19 - 20 2746 95
AF267981.1 - 19 - 20 2764 95

View Full Product Details