The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270364.1 | NP_001257293.1 | 12 - 13 | 1719 | 104 |
NM_001270365.1 | NP_001257294.1 | 10 - 11 | 1518 | 104 | |
NM_012190.3 | NP_036322.2 | 12 - 13 | 1821 | 104 | |
XM_006713481.2 | XP_006713544.1 | 12 - 13 | 1560 | 104 | |
XM_011512355.1 | XP_011510657.1 | 12 - 13 | 1553 | 104 | |
GenBank mRNA | AB209299.1 | - | 12 - 13 | 1628 | 104 |
AK222535.1 | - | 12 - 13 | 1603 | 104 | |
AK222694.1 | - | 12 - 13 | 1603 | 104 | |
AK294392.1 | - | 10 - 11 | 1302 | 104 | |
AK308112.1 | - | 12 - 13 | 1560 | 104 | |
CR749807.1 | - | 12 - 13 | 1719 | 104 | |
KU178541.1 | - | 12 - 13 | 1655 | 104 | |
KU178542.1 | - | 11 - 12 | 1471 | 104 |