The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270364.1 | NP_001257293.1 | 18 - 19 | 2326 | 80 |
NM_001270365.1 | NP_001257294.1 | 16 - 17 | 2125 | 80 | |
NM_012190.3 | NP_036322.2 | 18 - 19 | 2428 | 80 | |
XM_006713481.2 | XP_006713544.1 | 18 - 19 | 2167 | 80 | |
XM_011512355.1 | XP_011510657.1 | 18 - 19 | 2160 | 80 | |
XM_017005613.1 | XP_016861102.1 | 16 - 17 | 2329 | 80 | |
XM_017005614.1 | XP_016861103.1 | 16 - 17 | 1945 | 80 | |
GenBank mRNA | AB209299.1 | - | 18 - 19 | 2235 | 80 |
AK124908.1 | - | 1 - 2 | 943 | 80 | |
AK222535.1 | - | 18 - 19 | 2210 | 80 | |
AK222694.1 | - | 18 - 19 | 2210 | 80 | |
AK294392.1 | - | 16 - 17 | 1909 | 80 | |
CR749807.1 | - | 18 - 19 | 2326 | 80 |