The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008697.1 | NP_001008697.1 | 10 - 11 | 1654 | 99 |
NM_012143.2 | NP_036275.1 | 9 - 10 | 1577 | 99 | |
XM_011530079.1 | XP_011528381.1 | 10 - 11 | 1637 | 99 | |
XM_017028735.1 | XP_016884224.1 | 10 - 11 | 1795 | 99 | |
GenBank mRNA | AK025443.1 | - | 8 - 9 | 1701 | 99 |
AK025739.1 | - | 5 - 6 | 1035 | 99 | |
AK298303.1 | - | 5 - 6 | 637 | 99 | |
AL050258.1 | - | 9 - 10 | 1590 | 99 | |
AL080147.1 | - | 9 - 10 | 1557 | 99 | |
BC011599.2 | - | 9 - 10 | 1554 | 99 | |
BC033080.1 | - | 9 - 10 | 1555 | 99 | |
BT007274.1 | - | 6 - 7 | 1327 | 99 | |
CT841511.2 | - | 7 - 8 | 1429 | 99 |