The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159322.1 | NP_001152794.1 | 1 - 2 | 366 | 138 |
NM_001159323.1 | NP_001152795.1 | 1 - 2 | 366 | 138 | |
NM_003706.2 | NP_003697.2 | 1 - 2 | 366 | 138 | |
XM_011527431.2 | XP_011525733.1 | 1 - 2 | 959 | 138 | |
XM_011527432.2 | XP_011525734.1 | 1 - 2 | 959 | 138 | |
XM_011527433.2 | XP_011525735.1 | 1 - 2 | 959 | 138 | |
XM_017027411.1 | XP_016882900.1 | 1 - 2 | 959 | 138 | |
XM_017027412.1 | XP_016882901.1 | 1 - 2 | 959 | 138 | |
XM_017027413.1 | XP_016882902.1 | 1 - 2 | 959 | 138 | |
XM_017027414.1 | XP_016882903.1 | 1 - 2 | 959 | 138 | |
GenBank mRNA | AF058921.1 | - | 1 - 2 | 277 | 138 |
AF065214.1 | - | 1 - 2 | 311 | 138 | |
AK022440.1 | - | 1 - 2 | 296 | 138 | |
AK295400.1 | - | 1 - 2 | 331 | 138 | |
AK314524.1 | - | 1 - 2 | 326 | 138 | |
BC017956.1 | - | 1 - 2 | 281 | 138 | |
BC063416.1 | - | 1 - 2 | 349 | 165 |