The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018489.2 | NP_060959.2 | 10 - 11 | 6807 | 76 |
XM_006711450.3 | XP_006711513.1 | 10 - 11 | 6498 | 76 | |
XM_006711451.3 | XP_006711514.1 | 11 - 12 | 6585 | 76 | |
XM_006711452.3 | XP_006711515.1 | 9 - 10 | 6627 | 76 | |
XM_011509770.2 | XP_011508072.1 | 9 - 10 | 2178 | 76 | |
XM_017001784.1 | XP_016857273.1 | 10 - 11 | 6584 | 76 | |
XM_017001785.1 | XP_016857274.1 | 10 - 11 | 6514 | 76 | |
XM_017001786.1 | XP_016857275.1 | 10 - 11 | 6807 | 76 | |
XM_017001788.1 | XP_016857277.1 | 7 - 8 | 1302 | 76 | |
GenBank mRNA | AF257305.1 | - | 10 - 11 | 6651 | 76 |