The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018489.2 | NP_060959.2 | 14 - 15 | 7443 | 132 |
XM_006711450.3 | XP_006711513.1 | 14 - 15 | 7134 | 132 | |
XM_006711451.3 | XP_006711514.1 | 15 - 16 | 7221 | 132 | |
XM_006711452.3 | XP_006711515.1 | 13 - 14 | 7263 | 132 | |
XM_011509770.2 | XP_011508072.1 | 13 - 14 | 2814 | 132 | |
XM_017001784.1 | XP_016857273.1 | 14 - 15 | 7220 | 132 | |
XM_017001785.1 | XP_016857274.1 | 14 - 15 | 7150 | 132 | |
XM_017001786.1 | XP_016857275.1 | 14 - 15 | 7443 | 132 | |
XM_017001788.1 | XP_016857277.1 | 11 - 12 | 1938 | 132 | |
GenBank mRNA | AF257305.1 | - | 14 - 15 | 7287 | 132 |