Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
Chromosome Location
Chr.17: 42810132 - 42824316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003766.4 NP_003757.1 1 - 2 185 63
XM_017025262.1 XP_016880751.1 1 - 2 180 63
XM_017025263.1 XP_016880752.1 1 - 2 180 63
XM_017025264.1 XP_016880753.1 1 - 2 180 63
GenBank mRNA AK223411.1 - 1 - 2 127 63
BC010276.1 - 1 - 2 132 63
DC296896.1 - 1 - 2 162 63
DC354547.1 - 1 - 2 185 63

View Full Product Details