Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
Chromosome Location
Chr.17: 42810132 - 42824316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313998.1 NP_001300927.1 7 - 8 843 71
NM_001313999.1 NP_001300928.1 6 - 7 681 71
NM_001314000.1 NP_001300929.1 5 - 6 615 71
NM_003766.4 NP_003757.1 7 - 8 865 71
XM_005257759.2 XP_005257816.1 7 - 8 787 71
XM_005257760.3 XP_005257817.1 7 - 8 787 71
XM_011525421.2 XP_011523723.1 7 - 8 787 71
XM_017025262.1 XP_016880751.1 7 - 8 860 71
XM_017025263.1 XP_016880752.1 7 - 8 860 71
XM_017025264.1 XP_016880753.1 7 - 8 860 71
GenBank mRNA AF077301.1 - 6 - 7 679 71
AF139131.1 - 7 - 8 799 71
AK223411.1 - 7 - 8 807 71
AK298619.1 - 5 - 6 587 71
AK299616.1 - 6 - 7 1487 71
AK312651.1 - 7 - 8 790 71
BC010276.1 - 7 - 8 812 71
BX647788.1 - 5 - 6 1582 71
KC776730.1 - 6 - 7 679 71
L38932.1 - 2 - 3 234 71

View Full Product Details