Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
Chromosome Location
Chr.17: 42810132 - 42824316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313998.1 NP_001300927.1 9 - 10 1139 107
NM_003766.4 NP_003757.1 9 - 10 1161 107
GenBank mRNA AF077301.1 - 8 - 9 975 107
AF139131.1 - 9 - 10 1095 107
AK223411.1 - 9 - 10 1103 107
AK299616.1 - 8 - 9 1783 107
BC010276.1 - 9 - 10 1108 107

View Full Product Details