The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014034.2 | NP_054753.1 | 1 - 2 | 445 | 78 |
GenBank mRNA | AB028628.1 | - | 1 - 2 | 271 | 78 |
AF151856.1 | - | 3 - 4 | 722 | 78 | |
AF161495.1 | - | 3 - 4 | 722 | 78 | |
AF279306.1 | - | 1 - 2 | 257 | 78 | |
AK025738.1 | - | 1 - 2 | 302 | 78 | |
AL050261.1 | - | 1 - 2 | 300 | 78 | |
BC010878.1 | - | 1 - 2 | 284 | 78 | |
CR457347.1 | - | 1 - 2 | 108 | 78 | |
DA902777.1 | - | 1 - 2 | 445 | 78 |