The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 12 - 13 | 1967 | 64 |
NM_001282754.1 | NP_001269683.1 | 12 - 13 | 1967 | 64 | |
XM_011545533.1 | XP_011543835.1 | 8 - 9 | 776 | 64 | |
XM_011545536.2 | XP_011543838.1 | 4 - 5 | 809 | 64 | |
XM_017029579.1 | XP_016885068.1 | 8 - 9 | 777 | 64 | |
GenBank mRNA | AD000712.1 | - | 12 - 13 | 1564 | 64 |
AK298030.1 | - | 12 - 13 | 1513 | 64 | |
AK301528.1 | - | 11 - 12 | 1703 | 64 | |
AK304551.1 | - | 5 - 6 | 804 | 64 | |
BC105057.1 | - | 12 - 13 | 1489 | 64 | |
BC105059.1 | - | 12 - 13 | 1489 | 64 | |
U60475.1 | - | 12 - 13 | 1398 | 64 | |
U75645.1 | - | 12 - 13 | 1481 | 64 | |
U82970.1 | - | 12 - 13 | 2008 | 64 | |
U87284.1 | - | 12 - 13 | 1606 | 64 |