Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR1
Entrez Gene ID
9948
Gene Name
WD repeat domain 1
Gene Aliases
AIP1, HEL-S-52, NORI-1
Chromosome Location
Chr.4: 10074339 - 10116983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005112.4 NP_005103.2 8 - 9 1153 58
NM_017491.3 NP_059830.1 11 - 12 1573 58
XM_017008880.1 XP_016864369.1 11 - 12 1766 58
GenBank mRNA AB010427.2 - 11 - 12 1492 58
AB209746.1 - 11 - 12 1346 58
AF020056.1 - 11 - 12 1401 58
AF020260.1 - 11 - 12 1401 58
AK127665.1 - 11 - 12 1313 58
AK222764.1 - 11 - 12 1445 58
AK222897.1 - 11 - 12 1477 58
AK225276.1 - 11 - 12 1445 58
AK291614.1 - 11 - 12 1423 58
AK302209.1 - 7 - 8 845 58
AL050108.1 - 9 - 10 1121 58
AL110208.1 - 2 - 3 175 58
BC000201.1 - 11 - 12 1372 58
BC002489.2 - 11 - 12 1364 58
BC030541.2 - 8 - 9 949 58
EU794675.1 - 11 - 12 1364 58

View Full Product Details