The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017491.3 | NP_059830.1 | 4 - 5 | 660 | 94 |
XM_017008880.1 | XP_016864369.1 | 4 - 5 | 694 | 94 | |
GenBank mRNA | AB010427.2 | - | 4 - 5 | 579 | 94 |
AB209746.1 | - | 4 - 5 | 433 | 94 | |
AF020056.1 | - | 4 - 5 | 488 | 94 | |
AF020260.1 | - | 4 - 5 | 488 | 94 | |
AF274954.1 | - | - | 415 | 95 | |
AK222764.1 | - | 4 - 5 | 532 | 94 | |
AK222897.1 | - | 4 - 5 | 564 | 94 | |
AK225276.1 | - | 4 - 5 | 532 | 94 | |
AK291614.1 | - | 4 - 5 | 510 | 94 | |
AL050108.1 | - | 2 - 3 | 208 | 94 | |
BC000201.1 | - | 4 - 5 | 459 | 94 | |
BC002489.2 | - | 4 - 5 | 451 | 94 | |
EU794675.1 | - | 4 - 5 | 451 | 94 |