The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005112.4 | NP_005103.2 | 4 - 5 | 574 | 117 |
NM_017491.3 | NP_059830.1 | 7 - 8 | 994 | 117 | |
GenBank mRNA | AB010427.2 | - | 7 - 8 | 913 | 117 |
AB209746.1 | - | 7 - 8 | 767 | 117 | |
AF020056.1 | - | 7 - 8 | 822 | 117 | |
AF020260.1 | - | 7 - 8 | 822 | 117 | |
AK127665.1 | - | 7 - 8 | 734 | 117 | |
AK222764.1 | - | 7 - 8 | 866 | 117 | |
AK222897.1 | - | 7 - 8 | 898 | 117 | |
AK225276.1 | - | 7 - 8 | 866 | 117 | |
AK291614.1 | - | 7 - 8 | 844 | 117 | |
AK299602.1 | - | 7 - 8 | 670 | 117 | |
AK302209.1 | - | 3 - 4 | 266 | 117 | |
AL050108.1 | - | 5 - 6 | 542 | 117 | |
BC000201.1 | - | 7 - 8 | 793 | 117 | |
BC002489.2 | - | 7 - 8 | 785 | 117 | |
BC030541.2 | - | 4 - 5 | 370 | 117 | |
EU794675.1 | - | 7 - 8 | 785 | 117 |