Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR1
Entrez Gene ID
9948
Gene Name
WD repeat domain 1
Gene Aliases
AIP1, HEL-S-52, NORI-1
Chromosome Location
Chr.4: 10074339 - 10116983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005112.4 NP_005103.2 5 - 6 820 74
NM_017491.3 NP_059830.1 8 - 9 1240 74
XM_017008880.1 XP_016864369.1 8 - 9 1433 74
GenBank mRNA AB010427.2 - 8 - 9 1159 74
AB209746.1 - 8 - 9 1013 74
AF020056.1 - 8 - 9 1068 74
AF020260.1 - 8 - 9 1068 74
AK127665.1 - 8 - 9 980 74
AK222764.1 - 8 - 9 1112 74
AK222897.1 - 8 - 9 1144 74
AK225276.1 - 8 - 9 1112 74
AK291614.1 - 8 - 9 1090 74
AK299602.1 - 8 - 9 916 74
AK302209.1 - 4 - 5 512 74
AK303132.1 - - 641 74
AL050108.1 - 6 - 7 788 74
BC000201.1 - 8 - 9 1039 74
BC002489.2 - 8 - 9 1031 74
BC030541.2 - 5 - 6 616 74
BX648190.1 - 4 - 5 1890 74
EU794675.1 - 8 - 9 1031 74

View Full Product Details