Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR1
Entrez Gene ID
9948
Gene Name
WD repeat domain 1
Gene Aliases
AIP1, HEL-S-52, NORI-1
Chromosome Location
Chr.4: 10074339 - 10116983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005112.4 NP_005103.2 6 - 7 902 88
NM_017491.3 NP_059830.1 9 - 10 1322 88
XM_017008880.1 XP_016864369.1 9 - 10 1515 88
GenBank mRNA AB010427.2 - 9 - 10 1241 88
AB209746.1 - 9 - 10 1095 88
AF020056.1 - 9 - 10 1150 88
AF020260.1 - 9 - 10 1150 88
AK127665.1 - 9 - 10 1062 88
AK222764.1 - 9 - 10 1194 88
AK222897.1 - 9 - 10 1226 88
AK225276.1 - 9 - 10 1194 88
AK291614.1 - 9 - 10 1172 88
AK299602.1 - 9 - 10 998 88
AK302209.1 - 5 - 6 594 88
AK303132.1 - - 723 88
AL050108.1 - 7 - 8 870 88
BC000201.1 - 9 - 10 1121 88
BC002489.2 - 9 - 10 1113 88
BC030541.2 - 6 - 7 698 88
BX648190.1 - 5 - 6 1972 88
EU794675.1 - 9 - 10 1113 88

View Full Product Details