Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEY2
Entrez Gene ID
23493
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Gene Aliases
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32
Chromosome Location
Chr.6: 125747592 - 125762243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012259.2 NP_036391.1 2 - 3 344 107
GenBank mRNA AB044755.1 - 2 - 3 249 107
AF173901.1 - 2 - 3 312 107
AF232238.1 - 2 - 3 263 107
AF237949.1 - 2 - 3 220 107
AF311884.1 - 2 - 3 153 107
AJ249545.1 - 2 - 3 203 107
AK289943.1 - 2 - 3 262 107
BC007707.2 - 2 - 3 242 107
BT020067.1 - 2 - 3 153 107
BT020068.1 - 2 - 3 153 107

View Full Product Details