The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014400.2 | NP_055215.2 | 3 - 4 | 492 | 73 |
GenBank mRNA | AF082889.1 | - | 3 - 4 | 469 | 73 |
AJ223603.2 | - | 3 - 4 | 420 | 73 | |
AK075464.1 | - | 3 - 4 | 421 | 73 | |
AK314773.1 | - | 3 - 4 | 421 | 73 | |
AY359006.1 | - | 3 - 4 | 399 | 73 | |
BC039167.1 | - | 3 - 4 | 406 | 73 | |
BC041061.1 | - | 3 - 4 | 401 | 73 | |
BP367221.1 | - | 3 - 4 | 466 | 73 | |
DA837818.1 | - | 3 - 4 | 492 | 73 |