Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
Chromosome Location
Chr.16: 77190919 - 77199646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286639.1 NP_001273568.1 3 - 4 1314 70
NM_001286640.1 NP_001273569.1 2 - 3 1156 70
NM_014940.3 NP_055755.1 4 - 5 1641 70
GenBank mRNA AB020679.1 - 4 - 5 1325 70
AF442486.1 - 3 - 4 1305 70
AK023896.1 - 4 - 5 1369 70
AK128411.1 - 1 - 2 1183 70
AK293392.1 - 2 - 3 876 70
AK296467.1 - 3 - 4 1041 70
BC024277.1 - 4 - 5 1621 70

View Full Product Details