The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201461.1 | NP_001188390.1 | 11 - 12 | 1393 | 57 |
NM_020659.3 | NP_065710.1 | 11 - 12 | 1393 | 57 | |
XM_017027026.1 | XP_016882515.1 | 11 - 12 | 1237 | 57 | |
GenBank mRNA | AB188496.1 | - | 11 - 12 | 1393 | 57 |
AF177909.1 | - | 11 - 12 | 1307 | 57 | |
AK293468.1 | - | 12 - 13 | 1424 | 57 | |
BC019358.1 | - | 8 - 9 | 726 | 57 |