Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr.19: 45815442 - 45863290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004819.2 NP_004810.2 10 - 11 1488 61
XM_005259286.1 XP_005259343.1 10 - 11 1441 61
XM_011527354.1 XP_011525656.1 11 - 12 1491 61
GenBank mRNA AB209024.1 - 10 - 11 3736 61
AB210010.1 - 10 - 11 1333 61
AK300252.1 - 10 - 11 1342 61
BC006536.2 - 9 - 10 1195 61
BC006567.2 - 9 - 10 1195 61
BC030214.1 - 10 - 11 1330 61
BX649165.1 - 10 - 11 1343 61
U49240.1 - 5 - 6 957 61
Y10931.1 - 10 - 11 1306 61

View Full Product Details