The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004819.2 | NP_004810.2 | 19 - 20 | 2849 | 63 |
XM_005259286.1 | XP_005259343.1 | 19 - 20 | 2802 | 63 | |
XM_011527354.1 | XP_011525656.1 | 20 - 21 | 2852 | 63 | |
GenBank mRNA | AB209024.1 | - | 19 - 20 | 5012 | 63 |
AB210010.1 | - | 19 - 20 | 2694 | 63 | |
AK130602.1 | - | 10 - 11 | 988 | 63 | |
AK300252.1 | - | 19 - 20 | 2703 | 63 | |
BC006536.2 | - | 18 - 19 | 2556 | 63 | |
BC006567.2 | - | 18 - 19 | 2556 | 63 | |
BX649165.1 | - | 18 - 19 | 3755 | 63 | |
U49240.1 | - | 14 - 15 | 2318 | 63 | |
Y10931.1 | - | 19 - 20 | 2667 | 63 |