Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr.19: 45815442 - 45863290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004819.2 NP_004810.2 21 - 22 3043 93
XM_005259286.1 XP_005259343.1 21 - 22 2996 93
XM_011527354.1 XP_011525656.1 22 - 23 3046 93
GenBank mRNA AB209024.1 - 21 - 22 5206 93
AB210010.1 - 21 - 22 2888 93
AK130602.1 - 12 - 13 1182 93
AK300252.1 - 21 - 22 2897 93
BC006536.2 - 20 - 21 2750 93
BC006567.2 - 20 - 21 2750 93
BX649165.1 - 20 - 21 3949 93
U49240.1 - 16 - 17 2512 93
Y10931.1 - 21 - 22 2861 93

View Full Product Details