Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr.19: 45815442 - 45863290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004819.2 NP_004810.2 24 - 25 3502 66
XM_005259286.1 XP_005259343.1 24 - 25 3455 66
XM_011527354.1 XP_011525656.1 25 - 26 3505 66
GenBank mRNA AB209024.1 - 24 - 25 5665 66
AK130602.1 - 15 - 16 1641 66
AK300252.1 - 24 - 25 3356 66
BC006536.2 - 23 - 24 3209 66
BC006567.2 - 23 - 24 3209 66
BX649165.1 - 23 - 24 4408 66
U49240.1 - 19 - 20 2971 66
Y10931.1 - 24 - 25 3320 66

View Full Product Details