The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021255.2 | NP_067078.1 | 4 - 5 | 798 | 95 |
XM_005267890.4 | XP_005267947.1 | 4 - 5 | 843 | 95 | |
XM_006720211.3 | XP_006720274.1 | 3 - 4 | 2933 | 95 | |
XM_011536990.2 | XP_011535292.1 | 5 - 6 | 508 | 95 | |
XM_011536992.2 | XP_011535294.1 | 4 - 5 | 561 | 95 | |
XM_017021478.1 | XP_016876967.1 | 4 - 5 | 1190 | 95 | |
XM_017021479.1 | XP_016876968.1 | 5 - 6 | 9740 | 95 | |
GenBank mRNA | AF302502.1 | - | 4 - 5 | 688 | 95 |
AK315727.1 | - | 4 - 5 | 760 | 95 | |
BC009476.2 | - | 4 - 5 | 798 | 95 |