Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A5
Entrez Gene ID
1287
Gene Name
collagen type IV alpha 5 chain
Gene Aliases
ASLN, ATS, CA54
Chromosome Location
Chr.X: 108439844 - 108697545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000495.4 NP_000486.1 33 - 34 3198 78
NM_033380.2 NP_203699.1 33 - 34 3198 78
XM_011530849.2 XP_011529151.2 34 - 35 2946 78
XM_017029259.1 XP_016884748.1 34 - 35 2946 78
XM_017029260.1 XP_016884749.1 34 - 35 2946 78
XM_017029261.1 XP_016884750.1 34 - 35 2946 78
XM_017029262.1 XP_016884751.1 34 - 35 2946 78
XM_017029263.1 XP_016884752.1 18 - 19 2024 78
GenBank mRNA BC035387.1 - 17 - 18 2373 78
BC151846.1 - 33 - 34 3160 78
KM282618.1 - 33 - 34 3198 78
M31115.1 - 2 - 3 177 78
M58526.1 - 30 - 31 2674 78

View Full Product Details