The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000495.4 | NP_000486.1 | 40 - 41 | 3883 | 84 |
NM_033380.2 | NP_203699.1 | 40 - 41 | 3883 | 84 | |
XM_011530849.2 | XP_011529151.2 | 41 - 42 | 3631 | 84 | |
XM_017029259.1 | XP_016884748.1 | 41 - 42 | 3631 | 84 | |
XM_017029260.1 | XP_016884749.1 | 41 - 42 | 3631 | 84 | |
XM_017029261.1 | XP_016884750.1 | 41 - 42 | 3631 | 84 | |
XM_017029262.1 | XP_016884751.1 | 41 - 42 | 3631 | 84 | |
XM_017029263.1 | XP_016884752.1 | 25 - 26 | 2709 | 84 | |
GenBank mRNA | BC151846.1 | - | 40 - 41 | 3845 | 84 |
KM282618.1 | - | 40 - 41 | 3883 | 84 | |
M31115.1 | - | 9 - 10 | 862 | 84 | |
M58526.1 | - | 37 - 38 | 3359 | 84 |