The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000495.4 | NP_000486.1 | 8 - 9 | 747 | 99 |
NM_033380.2 | NP_203699.1 | 8 - 9 | 747 | 99 | |
XM_011530849.2 | XP_011529151.2 | 9 - 10 | 495 | 99 | |
XM_017029259.1 | XP_016884748.1 | 9 - 10 | 495 | 99 | |
XM_017029260.1 | XP_016884749.1 | 9 - 10 | 495 | 99 | |
XM_017029261.1 | XP_016884750.1 | 9 - 10 | 495 | 99 | |
XM_017029262.1 | XP_016884751.1 | 9 - 10 | 495 | 99 | |
GenBank mRNA | BC151846.1 | - | 8 - 9 | 709 | 99 |
KM282618.1 | - | 8 - 9 | 747 | 99 | |
M58526.1 | - | 5 - 6 | 223 | 99 | |
M90464.1 | - | 8 - 9 | 667 | 99 |