The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001961.3 | NP_001952.1 | 3 - 4 | 481 | 72 |
GenBank mRNA | AK297398.1 | - | 3 - 4 | 481 | 72 |
AK298498.1 | - | 3 - 4 | 481 | 72 | |
AK299225.1 | - | 3 - 4 | 477 | 72 | |
AU118535.1 | - | 3 - 4 | 481 | 72 | |
AY942181.1 | - | 2 - 3 | 398 | 72 | |
BC006547.2 | - | 3 - 4 | 466 | 72 | |
BC126259.1 | - | 3 - 4 | 450 | 72 | |
BC136313.1 | - | 3 - 4 | 450 | 72 | |
X51466.1 | - | 2 - 3 | 398 | 72 | |
Z11692.1 | - | 2 - 3 | 398 | 72 |