The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005253033.4 | XP_005253090.2 | 12 - 13 | 1517 | 122 |
XM_017018080.1 | XP_016873569.1 | 11 - 12 | 1394 | 122 | |
XM_017018081.1 | XP_016873570.1 | 11 - 12 | 1394 | 122 |