The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170690.1 | NP_001164161.1 | 17 - 18 | 2339 | 81 |
NM_020974.2 | NP_066025.2 | 21 - 22 | 2831 | 81 | |
XM_005253032.4 | XP_005253089.2 | 22 - 23 | 2998 | 81 | |
XM_005253033.4 | XP_005253090.2 | 21 - 22 | 2911 | 81 | |
XM_005253034.4 | XP_005253091.2 | 21 - 22 | 2875 | 81 | |
XM_005253036.4 | XP_005253093.2 | 19 - 20 | 2671 | 81 | |
XM_005253037.4 | XP_005253094.2 | 18 - 19 | 2533 | 81 | |
XM_011520246.2 | XP_011518548.1 | 21 - 22 | 2800 | 81 | |
XM_011520247.2 | XP_011518549.1 | 20 - 21 | 2794 | 81 | |
XM_017018080.1 | XP_016873569.1 | 20 - 21 | 2788 | 81 | |
XM_017018081.1 | XP_016873570.1 | 19 - 20 | 2590 | 81 | |
GenBank mRNA | AK092153.1 | - | 2 - 3 | 1558 | 81 |
AK092553.1 | - | 9 - 10 | 1362 | 81 | |
AK123039.1 | - | 21 - 22 | 2827 | 81 | |
AK131552.1 | - | 17 - 18 | 2316 | 81 | |
BC111690.1 | - | 17 - 18 | 2282 | 81 |