The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003948.3 | NP_003939.1 | 1 - 2 | 488 | 153 |
XM_005263335.2 | XP_005263392.1 | 1 - 2 | 207 | 153 | |
XM_006714406.2 | XP_006714469.1 | 1 - 2 | 207 | 153 | |
XM_017008809.1 | XP_016864298.1 | 1 - 2 | 207 | 153 | |
XM_017008810.1 | XP_016864299.1 | 1 - 2 | 207 | 153 | |
XM_017008811.1 | XP_016864300.1 | 1 - 2 | 207 | 153 | |
GenBank mRNA | AK294866.1 | - | 1 - 2 | 474 | 153 |
AK312490.1 | - | 1 - 2 | 479 | 153 | |
U35146.1 | - | 1 - 2 | 338 | 153 |