The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317231.1 | NP_001304160.1 | 3 - 4 | 470 | 73 |
NM_001317233.1 | NP_001304162.1 | 3 - 4 | 470 | 73 | |
NM_014342.3 | NP_055157.1 | 3 - 4 | 470 | 73 | |
XM_006718172.2 | XP_006718235.1 | 3 - 4 | 413 | 73 | |
XM_011519959.2 | XP_011518261.1 | 3 - 4 | 413 | 73 | |
XM_011519961.2 | XP_011518263.1 | 3 - 4 | 413 | 73 | |
XM_017017463.1 | XP_016872952.1 | 3 - 4 | 389 | 73 | |
GenBank mRNA | AF085361.1 | - | 3 - 4 | 329 | 73 |
AF176008.1 | - | 3 - 4 | 330 | 73 | |
AK295218.1 | - | 3 - 4 | 350 | 73 | |
AK313032.1 | - | 3 - 4 | 413 | 73 | |
AY380792.1 | - | 3 - 4 | 328 | 73 | |
BC000875.1 | - | 3 - 4 | 326 | 73 |