The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024641.3 | NP_078917.2 | 1 - 2 | 139 | 87 |
GenBank mRNA | AK291940.1 | - | 1 - 2 | 70 | 87 |
AY048774.1 | - | 1 - 2 | 94 | 87 | |
AY048775.1 | - | 1 - 2 | 94 | 87 | |
BC146671.1 | - | 1 - 2 | 74 | 87 | |
BC150199.1 | - | 1 - 2 | 74 | 87 | |
BX537398.1 | - | 1 - 2 | 70 | 87 | |
BX640869.1 | - | 1 - 2 | 100 | 87 | |
CR936827.1 | - | 1 - 2 | 74 | 87 |