Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPT1
Entrez Gene ID
85465
Gene Name
ethanolaminephosphotransferase 1
Gene Aliases
SELI, SEPI
Chromosome Location
Chr.2: 26346086 - 26395891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB051511.1 - 5 - 6 682 97
AK095620.1 - 5 - 6 692 97
BC021229.2 - 5 - 6 637 97

View Full Product Details