Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPT1
Entrez Gene ID
85465
Gene Name
ethanolaminephosphotransferase 1
Gene Aliases
SELI, SEPI
Chromosome Location
Chr.2: 26346086 - 26395891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB051511.1 - 6 - 7 793 169
AK095620.1 - 6 - 7 803 169

View Full Product Details