The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182543.3 | NP_872349.1 | 4 - 5 | 852 | 156 |
XM_011519383.1 | XP_011517685.1 | 4 - 5 | 839 | 156 | |
XM_011519384.1 | XP_011517686.1 | 6 - 7 | 824 | 156 | |
XM_011519385.1 | XP_011517687.1 | 5 - 6 | 513 | 156 | |
XM_011519386.2 | XP_011517688.1 | 5 - 6 | 566 | 156 | |
XM_011519387.2 | XP_011517689.1 | 3 - 4 | 4077 | 156 | |
XM_011519389.2 | XP_011517691.1 | 2 - 3 | 191 | 156 | |
XM_017015906.1 | XP_016871395.1 | 4 - 5 | 839 | 156 | |
XM_017015907.1 | XP_016871396.1 | 5 - 6 | 749 | 156 | |
XM_017015908.1 | XP_016871397.1 | 5 - 6 | 566 | 156 | |
XM_017015909.1 | XP_016871398.1 | 3 - 4 | 4077 | 156 | |
XM_017015910.1 | XP_016871399.1 | 3 - 4 | 4153 | 156 | |
XM_017015911.1 | XP_016871400.1 | 3 - 4 | 4152 | 156 | |
XM_017015912.1 | XP_016871401.1 | 2 - 3 | 277 | 156 | |
XM_017015913.1 | XP_016871402.1 | 5 - 6 | 4304 | 156 | |
XM_017015914.1 | XP_016871403.1 | 2 - 3 | 3996 | 156 | |
GenBank mRNA | AK074323.1 | - | 4 - 5 | 805 | 156 |
BC035778.1 | - | 4 - 5 | 838 | 156 |