The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308295.1 | NP_001295224.1 | 6 - 7 | 746 | 155 |
NM_004808.2 | NP_004799.1 | 5 - 6 | 689 | 155 | |
XM_005252642.4 | XP_005252699.1 | 5 - 6 | 725 | 155 | |
XM_006717539.3 | XP_006717602.1 | 5 - 6 | 697 | 155 | |
XM_017016947.1 | XP_016872436.1 | 5 - 6 | 725 | 155 | |
XM_017016948.1 | XP_016872437.1 | 6 - 7 | 664 | 155 | |
XM_017016949.1 | XP_016872438.1 | 6 - 7 | 664 | 155 | |
XM_017016950.1 | XP_016872439.1 | 5 - 6 | 697 | 155 | |
XM_017016951.1 | XP_016872440.1 | 6 - 7 | 664 | 155 | |
XM_017016952.1 | XP_016872441.1 | 6 - 7 | 885 | 155 | |
XM_017016953.1 | XP_016872442.1 | 6 - 7 | 885 | 155 | |
GenBank mRNA | AF043325.1 | - | 5 - 6 | 651 | 155 |
AK094904.1 | - | 6 - 7 | 664 | 155 | |
AK295277.1 | - | 6 - 7 | 625 | 155 | |
AK315085.1 | - | 5 - 6 | 617 | 155 | |
BC006376.2 | - | 5 - 6 | 607 | 155 | |
BT007024.1 | - | 5 - 6 | 605 | 155 |